Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01794047_g1
          See other PKA-R1 GE Assays ›
          Gene Symbol:
          Pka-R1
          Gene Name:
          Protein kinase, cAMP-dependent, regulatory subunit type 1
          Gene Aliases:
          18304, CG12452, CG18677, CG3263, CG42341, CdkR, DRI, Dmel\CG42341, Dmel_CG42341, PKA, PKA R1, PKA-R1, PKA-RI, PKAR, PKARI, PKAi, PKAr, PKa-R1, Pka-RI, PkaR, RI, dPKA-RI, dr[[1]], pKA, pkA, pka-R1
          Chromosome Location:
          Chr.3L: 20816118 - 20836048 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0259243
          Assay ID Dm01794047_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pka-R1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014593.3 NP_001014593.1 2 - 3 1072 67
          NM_001014594.3 NP_001014594.1 2 - 3 1072 67
          NM_001014595.3 NP_001014595.1 3 - 4 1007 67
          NM_001014596.3 NP_001014596.1 3 - 4 677 67
          NM_001014597.2 NP_001014597.1 2 - 3 505 67
          NM_001014598.2 NP_001014598.1 2 - 3 752 67
          NM_001202215.1 NP_001189144.1 3 - 4 628 67
          NM_001202216.1 NP_001189145.1 2 - 3 679 67
          NM_001202217.1 NP_001189146.1 2 - 3 752 67
          NM_001202218.2 NP_001189147.1 2 - 3 1072 67
          NM_001202219.2 NP_001189148.1 3 - 4 1014 67
          NM_001202220.2 NP_001189149.1 3 - 4 677 67
          NM_001202221.1 NP_001189150.1 2 - 3 861 67
          NM_079465.4 NP_524189.2 2 - 3 679 67
          NM_141006.4 NP_649263.2 2 - 3 861 67
          NM_168875.4 NP_730573.2 2 - 3 1072 67
          NM_168876.6 NP_730574.2 3 - 4 1014 67
          NM_168877.2 NP_730576.1 3 - 4 628 67
          GenBank mRNA AY058709.1 - 2 - 3 1038 67
          AY122131.1 - 2 - 3 679 67
          BT133037.1 - 3 - 4 628 67

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein phosphorylation
          actin filament organization
          neuromuscular synaptic transmission
          ventral cord development
          molting cycle, chitin-based cuticle
          oocyte microtubule cytoskeleton polarization
          olfactory learning
          regulation of protein kinase A signaling
          cAMP-mediated signaling
          regulation of cAMP-mediated signaling
          response to ethanol
          negative regulation of smoothened signaling pathway
          positive regulation of smoothened signaling pathway
          positive regulation of feeding behavior
          cAMP-dependent protein kinase regulator activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline