Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01794048_g1
          See other PKA-R1 GE Assays ›
          Gene Symbol:
          Pka-R1
          Gene Name:
          Protein kinase, cAMP-dependent, regulatory subunit type 1
          Gene Aliases:
          18304, CG12452, CG18677, CG3263, CG42341, CdkR, DRI, Dmel\CG42341, Dmel_CG42341, PKA, PKA R1, PKA-R1, PKA-RI, PKAR, PKARI, PKAi, PKAr, PKa-R1, Pka-RI, PkaR, RI, dPKA-RI, dr[[1]], pKA, pkA, pka-R1
          Chromosome Location:
          Chr.3L: 20816118 - 20836048 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0259243
          Assay ID Dm01794048_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pka-R1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014593.3 NP_001014593.1 3 - 4 1451 118
          NM_001014594.3 NP_001014594.1 3 - 4 1451 118
          NM_001014595.3 NP_001014595.1 4 - 5 1386 118
          NM_001014596.3 NP_001014596.1 4 - 5 1056 118
          NM_001014597.2 NP_001014597.1 3 - 4 884 118
          NM_001014598.2 NP_001014598.1 3 - 4 1131 118
          NM_079465.4 NP_524189.2 3 - 4 1058 118
          NM_141006.4 NP_649263.2 3 - 4 1240 118
          NM_168875.4 NP_730573.2 3 - 4 1451 118
          NM_168876.6 NP_730574.2 4 - 5 1393 118
          NM_168877.2 NP_730576.1 4 - 5 1007 118
          GenBank mRNA AY058709.1 - 3 - 4 1417 118
          AY122131.1 - 3 - 4 1058 118

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          protein-binding activity modulator kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein phosphorylation
          actin filament organization
          neuromuscular synaptic transmission
          ventral cord development
          molting cycle, chitin-based cuticle
          oocyte microtubule cytoskeleton polarization
          olfactory learning
          regulation of protein kinase A signaling
          cAMP-mediated signaling
          regulation of cAMP-mediated signaling
          response to ethanol
          negative regulation of smoothened signaling pathway
          positive regulation of smoothened signaling pathway
          positive regulation of feeding behavior
          cAMP-dependent protein kinase regulator activity

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline