Search
Search

Galphao| Gene Symbol: Galphao | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_078960.5 | NP_523684.2 | 4 - 5 | 841 | 66 | |
| NM_165772.4 | NP_724934.1 | 4 - 5 | 723 | 66 | ||
| NM_165773.3 | NP_724935.1 | 5 - 6 | 778 | 66 | ||
| NM_176124.3 | NP_788304.1 | 4 - 5 | 650 | 66 | ||
| NM_176125.3 | NP_788305.1 | 4 - 5 | 705 | 66 | ||
| NM_176126.3 | NP_788306.1 | 4 - 5 | 623 | 66 | ||
| NM_176127.3 | NP_788307.1 | 4 - 5 | 733 | 66 | ||
| NM_206079.2 | NP_995801.1 | 3 - 4 | 853 | 66 | ||
| NM_206080.2 | NP_995802.1 | 3 - 4 | 605 | 66 | ||
| GenBank mRNA | AY121631.1 | - | 5 - 6 | 759 | 66 | |
| BT046134.2 | - | 4 - 5 | 858 | 66 | ||
| M29602.1 | - | 3 - 4 | 1618 | 66 | ||
| M29731.1 | - | 4 - 5 | 627 | 66 | ||
| M29732.1 | - | 4 - 5 | 981 | 66 | ||
| M30151.1 | - | 4 - 5 | 651 | 66 | ||
| M86660.1 | - | 3 - 4 | 716 | 66 | ||
Set Membership: |
|