Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01794618_g1
          See other LIS-1 GE Assays ›
          Gene Symbol:
          Lis-1
          Gene Name:
          Lissencephaly-1
          Gene Aliases:
          CG8440, D-Lis1, DLIS-1, DLIS1, DLis-1, DLis1, Dlis-1, Dlis1, DmLIS1, Dmel\CG8440, Dmel_CG8440, LIS-1, Lis, Lis 1, Lis1, dLis1, l(2)k13209, l(2R)W8, lis-1, lis1, n(2)k11702
          Chromosome Location:
          Chr.2R: 16178826 - 16185103 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0015754
          Assay ID Dm01794618_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lis-1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001259432.2 NP_001246361.1 6 - 7 935 76
          NM_057812.5 NP_477160.1 6 - 7 1010 76
          NM_176190.2 NP_788370.1 6 - 7 923 76
          NM_206130.2 NP_995852.1 6 - 7 1440 76
          GenBank mRNA AF117606.1 - 6 - 7 1014 76
          AF152419.1 - 6 - 7 999 76
          BT021404.1 - 6 - 7 1005 76

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Drosophila melanogaster

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          intracellular protein transport
          mitotic cell cycle checkpoint
          nuclear migration
          germ cell development
          spermatogenesis
          germarium-derived oocyte fate determination
          border follicle cell migration
          cytoplasmic transport, nurse cell to oocyte
          oocyte nucleus migration involved in oocyte dorsal/ventral axis specification
          neuroblast proliferation
          axo-dendritic transport
          retrograde axonal transport
          protein localization
          intracellular mRNA localization
          transport along microtubule
          mushroom body development
          dendrite development
          stem cell population maintenance
          chorion-containing eggshell pattern formation
          nuclear migration along microtubule
          positive regulation of BMP signaling pathway
          germarium-derived oocyte differentiation
          oocyte fate determination
          ovarian fusome organization
          microtubule organizing center organization
          protein localization to kinetochore
          rhabdomere development
          regulation of translation by machinery localization
          fusome organization
          muscle cell cellular homeostasis
          dorsal appendage formation
          vesicle transport along microtubule
          female germ-line cyst formation
          germarium-derived cystoblast division
          oogenesis
          dendrite morphogenesis
          regulation of dendrite morphogenesis
          brain morphogenesis
          positive regulation of axonogenesis
          spindle assembly
          centrosome separation
          kinetochore organization
          centrosome localization
          nucleus localization
          clathrin-dependent endocytosis involved in vitellogenesis
          photoreceptor cell axon guidance
          axonal transport
          regulation of microtubule motor activity
          microtubule plus-end binding
          dynein complex binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline