Search
Search

smid| Gene Symbol: smid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274583.1 | NP_001261512.1 | 3 - 4 | 775 | 76 | |
| NM_079235.4 | NP_523959.2 | 3 - 4 | 632 | 76 | ||
| NM_206287.3 | NP_996009.2 | 3 - 4 | 542 | 76 | ||
| GenBank mRNA | BT124778.1 | - | 3 - 4 | 773 | 76 | |
Set Membership: |
|