Search
Search

lqf| Gene Symbol: lqf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274597.1 | NP_001261526.1 | 8 - 9 | 2718 | 56 | |
| NM_001274599.1 | NP_001261528.1 | 9 - 10 | 2875 | 56 | ||
| NM_001274600.1 | NP_001261529.1 | 7 - 8 | 2540 | 56 | ||
| NM_079238.3 | NP_523962.2 | 8 - 9 | 2838 | 56 | ||
| NM_168219.1 | NP_729266.2 | 9 - 10 | 2773 | 56 | ||
| NM_168220.1 | NP_729267.1 | 7 - 8 | 2660 | 56 | ||
| NM_168221.3 | NP_729268.2 | 8 - 9 | 2718 | 56 | ||
| NM_206291.2 | NP_996013.1 | 9 - 10 | 2995 | 56 | ||
| GenBank mRNA | BT133163.1 | - | 9 - 10 | 2981 | 56 | |
Set Membership: |
|