Search
Search

kcc| Gene Symbol: kcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001299864.1 | NP_001286793.1 | 4 - 5 | 1099 | 71 | |
| NM_001299865.1 | NP_001286794.1 | 3 - 4 | 557 | 71 | ||
| NM_001299866.1 | NP_001286795.1 | 3 - 4 | 566 | 71 | ||
| NM_131901.3 | NP_571976.2 | 3 - 4 | 489 | 71 | ||
| NM_166630.2 | NP_726377.1 | 3 - 4 | 828 | 71 | ||
| NM_166631.2 | NP_726378.1 | 3 - 4 | 581 | 71 | ||
| NM_166632.2 | NP_726379.1 | 3 - 4 | 566 | 71 | ||
| GenBank mRNA | AY118761.1 | - | 3 - 4 | 616 | 71 | |
| BT012491.1 | - | 3 - 4 | 851 | 71 | ||
Set Membership: |
|