Search
Search

cg| Gene Symbol: cg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103834.4 | NP_001097304.2 | 7 - 8 | 1988 | 70 | |
| NM_001103835.2 | NP_001097305.1 | 7 - 8 | 1964 | 70 | ||
| NM_001103836.4 | NP_001097306.2 | 7 - 8 | 2024 | 70 | ||
| NM_001144194.2 | NP_001137666.1 | 7 - 8 | 1988 | 70 | ||
| NM_001299479.1 | NP_001286408.1 | 6 - 7 | 1664 | 70 | ||
| NM_001299480.1 | NP_001286409.1 | 7 - 8 | 2009 | 70 | ||
| NM_001316419.1 | NP_001303348.1 | 6 - 7 | 1709 | 70 | ||
| NM_079014.3 | NP_523738.2 | 6 - 7 | 1685 | 70 | ||
| NM_166034.2 | NP_725362.1 | 7 - 8 | 1988 | 70 | ||
| NM_166035.2 | NP_725363.1 | 7 - 8 | 1508 | 70 | ||
| NM_166036.2 | NP_725364.1 | 7 - 8 | 1493 | 70 | ||
| GenBank mRNA | AF276664.1 | - | 6 - 7 | 1704 | 70 | |
| BT023511.1 | - | 6 - 7 | 1700 | 70 | ||
Set Membership: |
|