Search
Search

Flo1| Gene Symbol: Flo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058010.4 | NP_477358.1 | 1 - 2 | 286 | 57 | |
| NM_166102.2 | NP_725476.1 | 1 - 2 | 286 | 57 | ||
| GenBank mRNA | AF044734.1 | - | 1 - 2 | 52 | 57 | |
| AY058794.1 | - | 1 - 2 | 261 | 57 | ||
Set Membership: |
|