Search
Search

Strn-Mlck| Gene Symbol: Strn-Mlck | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202020.1 | NP_001188949.1 | 2 - 3 | 306 | 87 | |
| NM_001202026.2 | NP_001188955.2 | 16 - 17 | 20521 | 87 | ||
| NM_001274089.1 | NP_001261018.1 | 16 - 17 | 20521 | 87 | ||
| NM_001274090.1 | NP_001261019.1 | 16 - 17 | 20889 | 87 | ||
| NM_166129.3 | NP_725510.2 | 16 - 17 | 20889 | 87 | ||
| GenBank mRNA | AY113646.1 | - | 2 - 3 | 307 | 87 | |
Set Membership: |
|