Search
Search

spin| Gene Symbol: spin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080084.3 | NP_524823.1 | 5 - 6 | 1850 | 62 | |
| NM_166144.2 | NP_725530.1 | 5 - 6 | 1850 | 62 | ||
| NM_166145.2 | NP_725531.1 | 5 - 6 | 1850 | 62 | ||
| NM_166146.2 | NP_725532.1 | 5 - 6 | 1850 | 62 | ||
| NM_166147.2 | NP_725533.1 | 4 - 5 | 1588 | 62 | ||
| GenBank mRNA | BT032842.1 | - | 5 - 6 | 1812 | 62 | |
| BT150099.1 | - | 5 - 6 | 1806 | 62 | ||
Set Membership: |
|