Search
Search

spin| Gene Symbol: spin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080084.3 | NP_524823.1 | 3 - 4 | 1489 | 60 | |
| NM_166145.2 | NP_725531.1 | 3 - 4 | 1489 | 60 | ||
| GenBank mRNA | AF212367.1 | - | 3 - 4 | 1452 | 60 | |
| AF212369.1 | - | 3 - 4 | 1452 | 60 | ||
| BT150099.1 | - | 3 - 4 | 1445 | 60 | ||
Set Membership: |
|