Search
Search

ORMDL| Gene Symbol: ORMDL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_168929.2 | NP_730669.1 | 1 - 2 | 261 | 58 | |
| GenBank mRNA | BT023364.1 | - | 1 - 2 | 239 | 58 | |
| BT029397.1 | - | 1 - 2 | 181 | 58 | ||
| KX531694.1 | - | 1 - 2 | 181 | 58 | ||
Set Membership: |
|