Search
Search

Arf79F| Gene Symbol: Arf79F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104197.3 | NP_001097667.1 | 5 - 6 | 752 | 67 | |
| NM_001104198.2 | NP_001097668.2 | 4 - 5 | 718 | 67 | ||
| NM_001275296.1 | NP_001262225.1 | 4 - 5 | 723 | 67 | ||
| NM_057607.6 | NP_476955.1 | 4 - 5 | 808 | 67 | ||
| NM_168970.4 | NP_730757.1 | 4 - 5 | 672 | 67 | ||
| NM_168971.4 | NP_730758.1 | 4 - 5 | 784 | 67 | ||
| NM_168972.5 | NP_730759.1 | 4 - 5 | 587 | 67 | ||
| NM_168973.3 | NP_730760.1 | 4 - 5 | 783 | 67 | ||
| GenBank mRNA | AY060375.1 | - | 4 - 5 | 601 | 67 | |
Set Membership: |
|