Search
Search

shd| Gene Symbol: shd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274914.1 | NP_001261843.1 | 4 - 5 | 1618 | 61 | |
| NM_140452.2 | NP_648709.2 | 2 - 3 | 626 | 61 | ||
| NM_206352.2 | NP_996074.1 | 3 - 4 | 1363 | 61 | ||
| GenBank mRNA | AY089309.1 | - | 2 - 3 | 626 | 61 | |
Set Membership: |
|