Search
Search

dlp| Gene Symbol: dlp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079347.3 | NP_524071.2 | 4 - 5 | 1346 | 61 | |
| NM_206353.2 | NP_996075.2 | 4 - 5 | 1346 | 61 | ||
| GenBank mRNA | AY052017.1 | - | 4 - 5 | 1327 | 61 | |
Set Membership: |
|