Search
Search

Lmpt| Gene Symbol: Lmpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259879.2 | NP_001246808.1 | 2 - 3 | 415 | 60 | |
| NM_140673.5 | NP_648930.1 | 2 - 3 | 426 | 60 | ||
| NM_206388.3 | NP_996110.1 | 2 - 3 | 538 | 60 | ||
| GenBank mRNA | AJ418713.1 | - | 2 - 3 | 408 | 60 | |
| AY069696.1 | - | 2 - 3 | 408 | 60 | ||
| BT009969.1 | - | 2 - 3 | 423 | 60 | ||
Set Membership: |
|