Search
Search

SK| Gene Symbol: SK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103415.2 | NP_001096885.2 | 8 - 9 | 1578 | 74 | |
| NM_001272322.1 | NP_001259251.1 | 11 - 12 | 3006 | 74 | ||
| NM_001272323.2 | NP_001259252.1 | 10 - 11 | 2904 | 74 | ||
| NM_001272324.2 | NP_001259253.1 | 10 - 11 | 2904 | 74 | ||
| NM_001297957.1 | NP_001284886.1 | 8 - 9 | 1823 | 74 | ||
| NM_001297958.1 | NP_001284887.1 | 10 - 11 | 2904 | 74 | ||
| NM_001297959.1 | NP_001284888.1 | 8 - 9 | 1817 | 74 | ||
| NM_001297960.1 | NP_001284889.1 | 10 - 11 | 2907 | 74 | ||
| NM_080339.3 | NP_525078.3 | 8 - 9 | 1817 | 74 | ||
| NM_167029.3 | NP_726986.3 | 10 - 11 | 2907 | 74 | ||
| NM_167030.2 | NP_726987.2 | 8 - 9 | 1817 | 74 | ||
| NM_167031.3 | NP_726988.3 | 10 - 11 | 2907 | 74 | ||
| NM_167032.2 | NP_726989.3 | 8 - 9 | 1823 | 74 | ||
| NM_206631.4 | NP_996354.2 | 10 - 11 | 2907 | 74 | ||
Set Membership: |
|