Search
Search

sdt| Gene Symbol: sdt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038746.3 | NP_001033835.2 | 14 - 15 | 6974 | 77 | |
| NM_001038747.3 | NP_001033836.1 | 9 - 10 | 2994 | 77 | ||
| NM_001042798.2 | NP_001036263.1 | 7 - 8 | 2529 | 77 | ||
| NM_001258646.1 | NP_001245575.1 | 5 - 6 | 2302 | 77 | ||
| NM_001272397.1 | NP_001259326.1 | 14 - 15 | 6617 | 77 | ||
| NM_001272398.1 | NP_001259327.1 | 9 - 10 | 4162 | 77 | ||
| NM_001298081.1 | NP_001285010.1 | 13 - 14 | 5732 | 77 | ||
| NM_001298082.1 | NP_001285011.1 | 12 - 13 | 4433 | 77 | ||
| NM_132235.5 | NP_572463.1 | 10 - 11 | 4293 | 77 | ||
| NM_132236.6 | NP_572464.4 | 9 - 10 | 3726 | 77 | ||
| NM_206652.2 | NP_996375.1 | 5 - 6 | 2350 | 77 | ||
| NM_206653.3 | NP_996376.1 | 9 - 10 | 3955 | 77 | ||
| GenBank mRNA | BT010281.1 | - | 9 - 10 | 2992 | 77 | |
| BT099747.1 | - | 5 - 6 | 2350 | 77 | ||
Set Membership: |
|