Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01799208_g1
          See other MOE GE Assays ›
          Gene Symbol:
          Moe
          Gene Name:
          Moesin
          Gene Aliases:
          CG10701, D17, DMoe, Dmel\CG10701, Dmel_CG10701, Dmoe, EMR1, ERM, Emr1, GMA, Moes, anon-WO03040301.155, anon-WO03040301.157, anon-WO03040301.159, anon-WO03040301.161, dMoe, gma, l(1)G0067, l(1)G0323, l(1)G0404, l(1)G0415, moe, moesin, moesin/ezrin/radixin homolog mRNA
          Chromosome Location:
          Chr.X: 8873012 - 8898332 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0011661
          Assay ID Dm01799208_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Moe
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001258654.1 NP_001245583.1 7 - 8 1970 55
          NM_001258655.1 NP_001245584.1 6 - 7 2045 55
          NM_001272432.1 NP_001259361.1 8 - 9 1901 55
          NM_080343.4 NP_525082.2 6 - 7 1909 55
          NM_167168.4 NP_727290.1 7 - 8 1979 55
          NM_167169.2 NP_727291.1 7 - 8 1870 55
          NM_167170.2 NP_727292.1 3 - 4 1544 55
          NM_206664.3 NP_996387.1 6 - 7 1918 55
          NM_206665.3 NP_996388.1 7 - 8 1799 55
          NM_206666.3 NP_996389.1 6 - 7 1753 55
          NM_206667.3 NP_996390.1 6 - 7 1812 55
          NM_206668.3 NP_996391.1 6 - 7 1825 55
          NM_206669.2 NP_996392.1 6 - 7 1800 55
          GenBank mRNA AY069855.2 - 6 - 7 1893 55
          BT099729.1 - 7 - 8 1970 55
          L38909.1 - 6 - 7 1736 55

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          actin or actin-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelium
          RNA export from nucleus
          cytoskeleton organization
          oocyte anterior/posterior axis specification
          pole plasm assembly
          pole plasm protein localization
          determination of left/right symmetry
          protein localization
          positive regulation of cell proliferation
          regulation of cell shape
          anterior/posterior pattern specification
          regulation of membrane potential in photoreceptor cell
          establishment or maintenance of polarity of larval imaginal disc epithelium
          actin filament-based process
          actin cytoskeleton organization
          cortical actin cytoskeleton organization
          regulation of actin cytoskeleton organization
          microtubule anchoring
          establishment or maintenance of apical/basal cell polarity
          lumen formation, open tracheal system
          rhabdomere development
          eye photoreceptor cell development
          cortical microtubule organization
          establishment or maintenance of epithelial cell apical/basal polarity
          rhabdomere membrane biogenesis
          positive regulation of endocytosis
          branching involved in open tracheal system development
          photoreceptor cell axon guidance
          positive regulation of mitotic spindle elongation
          actin binding
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          microtubule binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline