Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01799269_m1
          See other DLG1 GE Assays ›
          Gene Symbol:
          dlg1
          Gene Name:
          discs large 1
          Gene Aliases:
          11, CG1725, CG1730, CPD, DLG, DLG-A, DLG1, Discs-large, Dlg, Dlg-A, Dlg1, DlgA, DlgS97, Dmel\CG1725, Dmel_CG1725, Drodlg, PSD-95, PSD95, SAP97, anon-EST:Posey93, anon-WO03040301.258, anon-WO03040301.260, anon-WO03040301.268, d. lg.-1, dlg, dlg-1, dlg-A, dlgA, dlgS97, l(1)10Bf, l(1)G0276, l(1)G0342, l(1)G0456, l(1)G19, l(1)L11, l(1)bwn, l(1)d.lg-1, l(1)d.lg.-1, l(1)discs large, l(1)dlg, l(1)dlg-1, l(1)dlg1, l(1)l.pr.-2, l(1)lpr-2, misb
          Chromosome Location:
          Chr.X: 11369665 - 11409776 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0001624
          Assay ID Dm01799269_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: dlg1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001103486.3 NP_001096956.1 3 - 4 956 113
          NM_001169247.1 NP_001162718.1 2 - 3 1083 113
          NM_001169249.3 NP_001162720.1 4 - 5 771 113
          NM_001272520.1 NP_001259449.1 3 - 4 956 113
          NM_167280.2 NP_727518.1 3 - 4 956 113
          NM_167281.2 NP_727519.1 2 - 3 1083 113
          NM_206679.2 NP_996402.1 3 - 4 504 113
          NM_206680.2 NP_996403.1 3 - 4 516 113
          NM_206681.4 NP_996404.1 3 - 4 956 113
          NM_206683.4 NP_996406.1 3 - 4 956 113
          GenBank mRNA AY059433.1 - 2 - 3 1083 113
          AY075410.1 - 3 - 4 948 113
          BT081965.1 - 4 - 5 743 113

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          cell fate specification
          morphogenesis of a polarized epithelium
          chemical synaptic transmission
          pole plasm protein localization
          dorsal closure
          nervous system development
          mating behavior
          male courtship behavior
          protein localization
          embryo development
          morphogenesis of follicular epithelium
          establishment or maintenance of polarity of follicular epithelium
          morphogenesis of larval imaginal disc epithelium
          establishment or maintenance of polarity of larval imaginal disc epithelium
          septate junction assembly
          ovarian follicle cell development
          anterior/posterior axis specification, follicular epithelium
          regulation of epidermal growth factor receptor signaling pathway
          gravitaxis
          receptor clustering
          asymmetric protein localization involved in cell fate determination
          basal protein localization
          establishment or maintenance of epithelial cell apical/basal polarity
          locomotor rhythm
          negative regulation of imaginal disc growth
          positive regulation of synaptic growth at neuromuscular junction
          positive phototaxis
          behavioral response to ethanol
          synaptic growth at neuromuscular junction
          establishment of spindle orientation
          cell fate commitment involved in pattern specification
          receptor localization to synapse
          cell-cell adhesion
          guanylate kinase activity
          protein binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline