Search
Search

AlkB| Gene Symbol: AlkB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347758.1 | NP_001334682.1 | 1 - 2 | 287 | 132 | |
| NM_206735.2 | NP_996458.1 | 1 - 2 | 179 | 132 | ||
| GenBank mRNA | BT030983.1 | - | 1 - 2 | 287 | 132 | |
Set Membership: |
|