Search
Search

Stim| Gene Symbol: Stim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258774.2 | NP_001245703.1 | 3 - 4 | 767 | 106 | |
| NM_078633.4 | NP_523357.2 | 3 - 4 | 703 | 106 | ||
| NM_206747.3 | NP_996470.2 | 3 - 4 | 703 | 106 | ||
| GenBank mRNA | AF328906.1 | - | 2 - 3 | 659 | 106 | |
| AY069686.1 | - | 3 - 4 | 687 | 106 | ||
| BT133089.1 | - | 3 - 4 | 681 | 106 | ||
Set Membership: |
|