Search
Search

nAChRalpha7| Gene Symbol: nAChRalpha7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169324.2 | NP_001162795.1 | 11 - 12 | 2566 | 111 | |
| NM_001298507.1 | NP_001285436.1 | 13 - 14 | 2549 | 111 | ||
| NM_167645.4 | NP_728239.3 | 12 - 13 | 2464 | 111 | ||
| NM_206791.3 | NP_996514.1 | 11 - 12 | 2566 | 111 | ||
Set Membership: |
|