Search
Search

CG17598| Gene Symbol: CG17598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272851.1 | NP_001259780.1 | 1 - 2 | 643 | 82 | |
| NM_134624.4 | NP_608468.1 | 1 - 2 | 643 | 82 | ||
| GenBank mRNA | AY070531.1 | - | 1 - 2 | 601 | 82 | |
Set Membership: |
|