Search
Search

dbr| Gene Symbol: dbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258880.2 | NP_001245809.1 | 3 - 4 | 793 | 77 | |
| NM_001258881.2 | NP_001245810.1 | 4 - 5 | 542 | 77 | ||
| NM_001258882.2 | NP_001245811.1 | 4 - 5 | 516 | 77 | ||
| NM_001258883.3 | NP_001245812.1 | 5 - 6 | 906 | 77 | ||
| NM_134642.3 | NP_608486.1 | 3 - 4 | 793 | 77 | ||
| GenBank mRNA | AF184228.1 | - | 3 - 4 | 760 | 77 | |
Set Membership: |
|