Search
Search

frtz| Gene Symbol: frtz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272950.1 | NP_001259879.1 | 1 - 2 | 710 | 65 | |
| NM_001272951.1 | NP_001259880.1 | 1 - 2 | 710 | 65 | ||
| NM_134770.2 | NP_608614.1 | 1 - 2 | 710 | 65 | ||
| GenBank mRNA | BT004832.1 | - | 1 - 2 | 722 | 65 | |
Set Membership: |
|