Search
Search

eys| Gene Symbol: eys | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032399.3 | NP_001027571.3 | 9 - 10 | 5588 | 66 | |
| NM_001272995.1 | NP_001259924.2 | 9 - 10 | 5588 | 66 | ||
| GenBank mRNA | DQ991915.1 | - | 9 - 10 | 5575 | 66 | |
Set Membership: |
|