Search
Search

Snx1| Gene Symbol: Snx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298657.1 | NP_001285586.1 | 1 - 2 | 298 | 60 | |
| NM_134933.2 | NP_608777.1 | 1 - 2 | 298 | 60 | ||
| GenBank mRNA | BT133048.1 | - | 1 - 2 | 295 | 60 | |
Set Membership: |
|