Search
Search

Art2| Gene Symbol: Art2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298671.1 | NP_001285600.1 | 3 - 4 | 619 | 97 | |
| NM_134977.2 | NP_608821.1 | 2 - 3 | 460 | 97 | ||
| GenBank mRNA | BT024987.1 | - | 2 - 3 | 460 | 97 | |
Set Membership: |
|