Search
Search

Tps1| Gene Symbol: Tps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134983.3 | NP_608827.1 | 4 - 5 | 2586 | 69 | |
| GenBank mRNA | AF145653.2 | - | 4 - 5 | 2548 | 69 | |
| BT021328.1 | - | 4 - 5 | 2592 | 69 | ||
Set Membership: |
|