Search
Search

path| Gene Symbol: path | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274705.1 | NP_001261634.1 | 5 - 6 | 758 | 66 | |
| NM_140070.3 | NP_648327.1 | 4 - 5 | 408 | 66 | ||
| NM_168349.2 | NP_729505.2 | 2 - 3 | 168 | 66 | ||
| GenBank mRNA | AY070669.1 | - | 4 - 5 | 410 | 66 | |
| AY070688.1 | - | 4 - 5 | 410 | 66 | ||
| BT088839.1 | - | 4 - 5 | 382 | 66 | ||
Set Membership: |
|