Search
Search

Ilp4| Gene Symbol: Ilp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_140104.3 | NP_648361.1 | 1 - 2 | 314 | 89 | |
| GenBank mRNA | AY071235.1 | - | 1 - 2 | 329 | 89 | |
| KX531430.1 | - | 1 - 2 | 164 | 89 | ||
Set Membership: |
|