Search
Search

simj| Gene Symbol: simj | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001274755.1 | NP_001261684.1 | 8 - 9 | 3258 | 86 | |
| NM_140150.3 | NP_648407.1 | 8 - 9 | 3222 | 86 | ||
| NM_168420.2 | NP_729624.1 | 8 - 9 | 3234 | 86 | ||
| NM_168421.2 | NP_729625.1 | 9 - 10 | 2993 | 86 | ||
| GenBank mRNA | AY051828.1 | - | 8 - 9 | 3187 | 86 | |
Set Membership: |
|