Search
Search

sip2| Gene Symbol: sip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135246.3 | NP_609090.2 | 2 - 3 | 1796 | 77 | |
| NM_164722.2 | NP_723234.2 | 3 - 4 | 1908 | 77 | ||
| NM_164723.1 | NP_723235.2 | 2 - 3 | 1704 | 77 | ||
| NM_164724.1 | NP_723236.1 | 2 - 3 | 1706 | 77 | ||
| NM_164725.3 | NP_723237.1 | 1 - 2 | 1725 | 77 | ||
| GenBank mRNA | BT031251.1 | - | 3 - 4 | 1845 | 77 | |
Set Membership: |
|