Search
Search

Oatp30B| Gene Symbol: Oatp30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135449.4 | NP_609293.2 | 5 - 6 | 2525 | 69 | |
| NM_164857.3 | NP_723463.1 | 5 - 6 | 2580 | 69 | ||
| NM_205945.2 | NP_995667.1 | 3 - 4 | 1921 | 69 | ||
| NM_205946.2 | NP_995668.1 | 4 - 5 | 2399 | 69 | ||
Set Membership: |
|