Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01803876_m1
          See other PKA-C1 GE Assays ›
          Gene Symbol:
          Pka-C1
          Gene Name:
          Protein kinase, cAMP-dependent, catalytic subunit 1
          Gene Aliases:
          6353, C, CG4379, CdkA, Cos, Cos-1, Cos1, DC0, DCO, Dc0, Dco, Dcpk, Dmel\CG4379, Dmel_CG4379, PKA, PKA CI, PKA Cl, PKA-C, PKA-C1, PKAC1, PKAc, PKAcA, Pka, Pka-C, PkaC1, cos1, dPKA, dc0, dcO, dco, l(2)01272, l(2)cos[[1]], l(2)s4402, pKA, pkA, pka, pka C1, pka-C1, pka-c1
          Chromosome Location:
          Chr.2L: 9682315 - 9699297 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0000273
          Assay ID Dm01803876_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pka-C1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_057629.4 NP_476977.1 2 - 3 2644 72
          NM_164866.3 NP_723479.1 2 - 3 2231 72
          NM_205950.3 NP_995672.1 2 - 3 2429 72
          GenBank mRNA AY069425.1 - 2 - 3 2217 72

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          G-protein coupled receptor signaling pathway
          oocyte anterior/posterior axis specification
          regulation of pole plasm oskar mRNA localization
          anterior/posterior pattern specification, imaginal disc
          imaginal disc-derived wing morphogenesis
          anesthesia-resistant memory
          rhythmic behavior
          oocyte microtubule cytoskeleton polarization
          olfactory learning
          regulation of bicoid mRNA localization
          positive regulation of gene expression
          protein kinase A signaling
          cAMP-mediated signaling
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of hippo signaling
          thermosensory behavior
          regulation of apoptotic process
          regulation of circadian sleep/wake cycle, sleep
          negative regulation of smoothened signaling pathway
          positive regulation of smoothened signaling pathway
          negative regulation of nucleocytoplasmic transport
          oogenesis
          sprouting of injured axon
          modulation of synaptic transmission
          cellular response to ethanol
          regulation of actin cytoskeleton reorganization
          protein serine/threonine kinase activity
          cAMP-dependent protein kinase activity
          protein binding
          ATP binding
          protein kinase A regulatory subunit binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline