Search
Search

JhI-21| Gene Symbol: JhI-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259067.1 | NP_001245996.1 | 5 - 6 | 947 | 73 | |
| NM_001298934.1 | NP_001285863.1 | 5 - 6 | 917 | 73 | ||
| NM_001298935.1 | NP_001285864.1 | 5 - 6 | 980 | 73 | ||
| NM_135698.4 | NP_609542.1 | 5 - 6 | 923 | 73 | ||
| NM_164988.4 | NP_723717.2 | 5 - 6 | 887 | 73 | ||
| GenBank mRNA | AF273478.1 | - | 5 - 6 | 885 | 73 | |
| AY051907.1 | - | 5 - 6 | 872 | 73 | ||
Set Membership: |
|