Search
Search

ThrRS| Gene Symbol: ThrRS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298939.1 | NP_001285868.1 | 3 - 4 | 808 | 84 | |
| NM_080100.6 | NP_524839.3 | 3 - 4 | 586 | 84 | ||
| NM_164994.3 | NP_723725.1 | 3 - 4 | 808 | 84 | ||
| NM_164995.2 | NP_723726.1 | 3 - 4 | 602 | 84 | ||
| GenBank mRNA | BT050469.1 | - | 3 - 4 | 765 | 84 | |
Set Membership: |
|