Search
Search

| Gene Symbol: Ir25a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273120.1 | NP_001260049.1 | 3 - 4 | 1014 | 101 | |
| NM_001273121.1 | NP_001260050.1 | 2 - 3 | 506 | 101 | ||
| NM_001273122.1 | NP_001260051.1 | 3 - 4 | 707 | 101 | ||
| NM_135019.2 | NP_608863.2 | 4 - 5 | 572 | 101 | ||
| GenBank mRNA | HQ600588.1 | - | 3 - 4 | 534 | 101 | |
| Gene Symbol: tank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT099717.1 | - | 7 - 8 | 2092 | 101 | |
Set Membership: |
|