Search
Search

bru1| Gene Symbol: bru1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001169492.2 | NP_001162963.1 | 3 - 4 | 991 | 70 | |
| NM_001273474.1 | NP_001260403.1 | 4 - 5 | 1629 | 70 | ||
| NM_001273475.1 | NP_001260404.1 | 3 - 4 | 863 | 70 | ||
| NM_001273476.1 | NP_001260405.1 | 3 - 4 | 716 | 70 | ||
| NM_001273477.1 | NP_001260406.1 | 3 - 4 | 1021 | 70 | ||
| NM_001273478.1 | NP_001260407.1 | 3 - 4 | 992 | 70 | ||
| NM_165002.2 | NP_723737.1 | 4 - 5 | 1776 | 70 | ||
| NM_165003.3 | NP_723738.2 | 3 - 4 | 991 | 70 | ||
| NM_165004.3 | NP_723739.1 | 3 - 4 | 991 | 70 | ||
| GenBank mRNA | AY051758.1 | - | 3 - 4 | 971 | 70 | |
| BT133212.1 | - | 4 - 5 | 1755 | 70 | ||
Set Membership: |
|