Search
Search

Pect| Gene Symbol: Pect | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_135769.3 | NP_609613.2 | 1 - 2 | 502 | 67 | |
| NM_165028.2 | NP_723789.2 | 1 - 2 | 502 | 67 | ||
| NM_165029.2 | NP_723790.2 | 1 - 2 | 502 | 67 | ||
| NM_165030.2 | NP_723791.2 | 1 - 2 | 502 | 67 | ||
| GenBank mRNA | BT003190.1 | - | 1 - 2 | 496 | 67 | |
| BT125823.1 | - | 1 - 2 | 213 | 67 | ||
Set Membership: |
|