Search
Search

yuri| Gene Symbol: yuri | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038821.2 | NP_001033910.2 | 7 - 8 | 1504 | 66 | |
| NM_001038822.3 | NP_001033911.3 | 8 - 9 | 1418 | 66 | ||
| NM_001316396.1 | NP_001303325.1 | 5 - 6 | 1295 | 66 | ||
| NM_165106.5 | NP_723903.3 | 10 - 11 | 2290 | 66 | ||
Set Membership: |
|