Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Dm01807836_g1
          See other MHC GE Assays ›
          Gene Symbol:
          Mhc
          Gene Name:
          Myosin heavy chain
          Gene Aliases:
          Bsh, CG17927, DROMHC, Dm II, DmMHC, Dmel\CG17927, Dmel_CG17927, DroMII, Ifm(2)2, Ifm(2)RU1, MHC, MRP, Mhc36B, Mrp, MyHC, Myo, Myo6, Nup, Sht, Stp, chr2L:16765388..16765564, chr2L:16781880..16782040, ifm(2)2, ifm(2)RU1, ifm(2)RU2, l(2)36Ae, l(2)M66, l(2)k10423, mMHC, mhc, sMHC
          Chromosome Location:
          Chr.2L: 16766566 - 16788766 on Build Release_6_plus_ISO1_MT
          Species:
          Drosophila melanogaster
          Species Specific ID (Flybase ID):
          FBgn0264695
          Assay ID Dm01807836_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mhc
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001169519.2 NP_001162990.1 4 - 5 851 64
          NM_001169520.3 NP_001162991.1 4 - 5 1013 64
          NM_001169521.2 NP_001162992.1 4 - 5 851 64
          NM_001259119.2 NP_001246048.1 4 - 5 851 64
          NM_001259120.1 NP_001246049.1 4 - 5 1006 64
          NM_001259121.2 NP_001246050.1 3 - 4 801 64
          NM_001259122.1 NP_001246051.1 4 - 5 1011 64
          NM_001316398.1 NP_001303327.1 4 - 5 1006 64
          NM_078863.6 NP_523587.4 4 - 5 851 64
          NM_165181.3 NP_723999.1 4 - 5 851 64
          NM_165182.3 NP_724000.1 4 - 5 851 64
          NM_165183.3 NP_724001.1 4 - 5 851 64
          NM_165184.3 NP_724002.2 4 - 5 993 64
          NM_165185.3 NP_724003.1 4 - 5 851 64
          NM_165186.3 NP_724004.1 4 - 5 851 64
          NM_165187.3 NP_724005.1 4 - 5 851 64
          NM_165188.3 NP_724006.1 4 - 5 851 64
          NM_165189.3 NP_724007.1 4 - 5 851 64
          NM_165190.4 NP_724008.1 4 - 5 1019 64
          NM_165191.3 NP_724009.1 4 - 5 851 64
          NM_165192.4 NP_724010.1 4 - 5 851 64

          Back To Top

          More Information


          Set Membership:

          Amplicon length less than or equal to 70 Drosophila melanogaster

          Gene Ontology Categories:

          Function(s) Process(es)

          muscle contraction
          border follicle cell migration
          epithelial cell migration, open tracheal system
          muscle organ development
          adult somatic muscle development
          myofibril assembly
          skeletal muscle myosin thick filament assembly
          myosin filament organization
          locomotion
          muscle cell differentiation
          regulation of myosin II filament assembly
          sarcomere organization
          protein stabilization
          flight
          muscle thin filament assembly
          motor activity
          ATP binding
          structural constituent of muscle
          actin-dependent ATPase activity
          protein homodimerization activity
          actin filament binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline