Search
Search

CG43340| Gene Symbol: CG43340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103740.3 | NP_001097210.2 | 9 - 10 | 1906 | 68 | |
| NM_001259255.1 | NP_001246184.1 | 9 - 10 | 1636 | 68 | ||
| NM_001259256.2 | NP_001246185.1 | 8 - 9 | 1845 | 68 | ||
| NM_001259257.1 | NP_001246186.1 | 9 - 10 | 1636 | 68 | ||
| NM_001259258.2 | NP_001246187.1 | 5 - 6 | 1213 | 68 | ||
| NM_001259259.2 | NP_001246188.1 | 5 - 6 | 1146 | 68 | ||
| NM_001259260.1 | NP_001246189.1 | 5 - 6 | 977 | 68 | ||
| NM_001273853.1 | NP_001260782.1 | 8 - 9 | 1550 | 68 | ||
| NM_001273854.1 | NP_001260783.1 | 3 - 4 | 438 | 68 | ||
| NM_136458.3 | NP_610302.3 | 9 - 10 | 1636 | 68 | ||
| NM_170613.3 | NP_724586.2 | 8 - 9 | 1550 | 68 | ||
| NM_170614.3 | NP_724587.2 | 8 - 9 | 1413 | 68 | ||
| NM_176100.2 | NP_788280.1 | 8 - 9 | 1816 | 68 | ||
| NM_176101.3 | NP_788281.1 | 8 - 9 | 1816 | 68 | ||
| NM_176102.3 | NP_788282.1 | 5 - 6 | 977 | 68 | ||
| GenBank mRNA | BT132651.1 | - | 8 - 9 | 1546 | 68 | |
Set Membership: |
|