Search
Search

mnb| Gene Symbol: mnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014750.2 | NP_001014750.1 | 4 - 5 | 2195 | 80 | |
| NM_001298464.1 | NP_001285393.1 | 3 - 4 | 916 | 80 | ||
| NM_167581.3 | NP_728104.1 | 4 - 5 | 2195 | 80 | ||
| NM_170658.3 | NP_728106.1 | 4 - 5 | 2195 | 80 | ||
| NM_170659.2 | NP_728107.2 | 3 - 4 | 916 | 80 | ||
Set Membership: |
|