Search
Search

retm| Gene Symbol: retm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001273203.2 | NP_001260132.1 | 5 - 6 | 2035 | 59 | |
| NM_001298743.1 | NP_001285672.1 | 5 - 6 | 2035 | 59 | ||
| NM_135184.4 | NP_609028.2 | 5 - 6 | 2035 | 59 | ||
| GenBank mRNA | BT044154.1 | - | 5 - 6 | 1970 | 59 | |
Set Membership: |
|