Search
Search

aust| Gene Symbol: aust | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001298846.1 | NP_001285775.1 | 1 - 2 | 292 | 79 | |
| NM_135434.2 | NP_609278.1 | 1 - 2 | 292 | 79 | ||
| GenBank mRNA | BT128872.1 | - | 1 - 2 | 292 | 79 | |
Set Membership: |
|