Search
Search

BicD| Gene Symbol: BicD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001259128.2 | NP_001246057.1 | 6 - 7 | 2945 | 63 | |
| NM_001273601.1 | NP_001260530.1 | 6 - 7 | 2945 | 63 | ||
| NM_001273602.1 | NP_001260531.1 | 6 - 7 | 2945 | 63 | ||
| NM_165220.3 | NP_724056.1 | 6 - 7 | 2945 | 63 | ||
| GenBank mRNA | AY069452.1 | - | 6 - 7 | 2448 | 63 | |
| X51652.1 | - | 2988 | 63 | |||
Set Membership: |
|