Search
Search

CadN| Gene Symbol: CadN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032108.2 | NP_001027279.1 | 15 - 16 | 5229 | 88 | |
| NM_001032109.3 | NP_001027280.1 | 15 - 16 | 5229 | 88 | ||
| NM_165224.2 | NP_724068.1 | 14 - 15 | 5217 | 88 | ||
| NM_165225.2 | NP_724069.1 | 14 - 15 | 5217 | 88 | ||
| NM_165228.2 | NP_724072.1 | 14 - 15 | 5217 | 88 | ||
| NM_165229.2 | NP_724073.1 | 14 - 15 | 5217 | 88 | ||
Set Membership: |
|